Assay Details
Gene Symbol: ARPC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336326.1 | NP_001323442.1 | 10 - 11 | 973 | 176 | |
NM_128686.3 | NP_180688.1 | 10 - 11 | 978 | 176 | ||
GenBank mRNA | AF439845.1 | - | 10 - 11 | 954 | 176 | |
BT000567.1 | - | 10 - 11 | 761 | 176 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
