Assay Details
Gene Symbol: HMGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035998.1 | NP_001031075.1 | 3 - 4 | 320 | 85 | |
NM_001332459.1 | NP_001322266.1 | 3 - 4 | 457 | 85 | ||
NM_101921.4 | NP_564124.1 | 3 - 4 | 488 | 85 | ||
GenBank mRNA | AF411789.1 | - | 3 - 4 | 204 | 85 | |
AY084848.1 | - | 3 - 4 | 295 | 85 | ||
AY097385.1 | - | 2 - 3 | 139 | 85 | ||
AY136309.1 | - | 3 - 4 | 259 | 85 | ||
BT000420.1 | - | 2 - 3 | 139 | 85 | ||
Y09482.1 | - | 187 | 85 | |||
Y14073.1 | - | 189 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
