Assay Details
Gene Symbol: MYB113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105308.2 | NP_176811.1 | 1 - 2 | 164 | 147 | |
GenBank mRNA | AY008378.1 | - | 1 - 2 | 119 | 147 | |
AY519566.1 | - | 1 - 2 | 119 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
