Assay Details
Gene Symbol: AT3G24130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113318.2 | NP_189055.1 | 2 - 3 | 441 | 150 | |
GenBank mRNA | DQ056606.1 | - | 2 - 3 | 378 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
