Assay Details
Gene Symbol: AT3G24130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113318.2 | NP_189055.1 | 3 - 4 | 658 | 139 | |
GenBank mRNA | DQ056606.1 | - | 3 - 4 | 595 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
