Assay Details
Gene Symbol: AT4G07950 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116865.3 | NP_192535.1 | 2 - 3 | 537 | 171 | |
GenBank mRNA | AY072451.1 | - | 2 - 3 | 334 | 171 | |
BT000318.1 | - | 1 - 2 | 203 | 171 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
