Assay Details
Gene Symbol: HSP81-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124983.4 | NP_200412.1 | 2 - 3 | 435 | 122 | |
GenBank mRNA | AY062832.1 | - | 2 - 3 | 368 | 122 | |
AY081302.1 | - | 2 - 3 | 366 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
