Assay Details
Gene Symbol: MYB116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036014.2 | NP_001031091.1 | 1 - 2 | 150 | 90 | |
NM_001332675.1 | NP_001321760.1 | 1 - 2 | 150 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
