Assay Details
Gene Symbol: ARPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036555.3 | NP_001031632.1 | 2 - 3 | 187 | 118 | |
NM_001203797.1 | NP_001190726.1 | 3 - 4 | 220 | 118 | ||
NM_001340903.1 | NP_001319932.1 | 3 - 4 | 405 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
