Assay Details
Gene Symbol: EDA31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111835.3 | NP_187611.2 | 1 - 2 | 459 | 80 | |
GenBank mRNA | AJ630487.1 | - | 1 - 2 | 367 | 80 | |
AY568659.1 | - | 1 - 2 | 367 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
