Assay Details
Gene Symbol: MYB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343462.1 | NP_001330269.1 | 2 - 3 | 500 | 149 | |
NM_001343463.1 | NP_001330270.1 | 1 - 2 | 704 | 149 | ||
NM_121683.4 | NP_197179.2 | 2 - 3 | 500 | 149 | ||
NM_203063.2 | NP_974792.1 | 1 - 2 | 754 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
