Assay Details
Gene Symbol: AT3G53010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115161.4 | NP_190869.3 | 1 - 2 | 801 | 143 | |
GenBank mRNA | AK229401.1 | - | 1 - 2 | 680 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
