Assay Details
Gene Symbol: AT4G12520 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117322.3 | NP_567392.1 | 1 | 535 | 151 |
Gene Symbol: AT4G12510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK227411.1 | - | 1 | 445 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
