Assay Details
Gene Symbol: ZNF148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033121.1 | NP_001028293.1 | 5 - 6 | 758 | 77 | |
GenBank mRNA | DQ162621.1 | - | 5 - 6 | 758 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
