Assay Details
Gene Symbol: PEG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002887.3 | NP_001002887.3 | 8 - 9 | 1203 | 80 | |
GenBank mRNA | AY427787.1 | - | 1135 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
