Assay Details
Gene Symbol: PTGES3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007806.2 | NP_001007807.2 | 4 - 5 | 480 | 142 | |
GenBank mRNA | AY692440.1 | - | 3 - 4 | 284 | 142 | |
BC103350.1 | - | 4 - 5 | 480 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
