Assay Details
Gene Symbol: PNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007818.1 | NP_001007819.1 | 4 - 5 | 557 | 90 | |
GenBank mRNA | AF529136.1 | - | 4 - 5 | 557 | 90 | |
BC103291.1 | - | 4 - 5 | 554 | 90 | ||
BT021545.1 | - | 4 - 5 | 553 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
