Assay Details
Gene Symbol: MPZL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015673.2 | NP_001015673.1 | 1 - 2 | 300 | 143 | |
GenBank mRNA | BC102486.1 | - | 1 - 2 | 192 | 143 | |
BT020659.1 | - | 1 - 2 | 301 | 143 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102486.1, NM_001015673.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
