Assay Details
Gene Symbol: PLVAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035353.2 | NP_001030430.1 | 5 - 6 | 1375 | 67 | |
GenBank mRNA | BC102834.1 | - | 5 - 6 | 1378 | 67 | |
BT025486.1 | - | 5 - 6 | 1326 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
