Assay Details
Gene Symbol: MAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075428.2 | NP_001068896.1 | 3 - 4 | 480 | 92 | |
GenBank mRNA | BC103042.1 | - | 3 - 4 | 481 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
