Assay Details
Gene Symbol: ELAVL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076454.2 | NP_001069922.1 | 5 - 6 | 829 | 100 | |
GenBank mRNA | BC102495.1 | - | 5 - 6 | 830 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
