Assay Details
Gene Symbol: F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173878.2 | NP_776303.1 | 2 - 3 | 354 | 91 | |
GenBank mRNA | BC123808.1 | - | 2 - 3 | 327 | 91 | |
S74147.1 | - | 2 - 3 | 354 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
