Assay Details
Gene Symbol: SERPINA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173882.2 | NP_776307.1 | 2 - 3 | 677 | 92 | |
GenBank mRNA | BC102730.1 | - | 2 - 3 | 695 | 92 | |
BT025459.1 | - | 2 - 3 | 787 | 92 | ||
X63129.1 | - | 678 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
