Assay Details
Gene Symbol: IL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173920.2 | NP_776345.1 | 2 - 3 | 213 | 144 | |
GenBank mRNA | L31893.1 | - | 2 - 3 | 213 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
