Assay Details
Gene Symbol: IL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173922.1 | NP_776347.1 | 3 - 4 | 307 | 144 | |
GenBank mRNA | EU276070.1 | - | 3 - 4 | 307 | 144 | |
Z67872.1 | - | 3 - 4 | 307 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
