Assay Details
Gene Symbol: MX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173940.2 | NP_776365.1 | 2 - 3 | 168 | 95 | |
GenBank mRNA | BC118109.1 | - | 2 - 3 | 157 | 95 | |
JQ766265.1 | - | 1 - 2 | 70 | 95 | ||
U88329.1 | - | 2 - 3 | 168 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
