Assay Details
Gene Symbol: MX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173940.2 | NP_776365.1 | 4 - 5 | 494 | 59 | |
GenBank mRNA | AB060169.1 | - | 3 - 4 | 870 | 59 | |
AF047692.1 | - | 3 - 4 | 416 | 59 | ||
BC118109.1 | - | 4 - 5 | 483 | 59 | ||
JQ766265.1 | - | 3 - 4 | 396 | 59 | ||
U88329.1 | - | 4 - 5 | 494 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
