Assay Details
Gene Symbol: MX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173940.2 | NP_776365.1 | 7 - 8 | 991 | 67 | |
GenBank mRNA | AB060169.1 | - | 6 - 7 | 1367 | 67 | |
AF047692.1 | - | 6 - 7 | 913 | 67 | ||
BC118109.1 | - | 7 - 8 | 980 | 67 | ||
JQ766265.1 | - | 6 - 7 | 893 | 67 | ||
U88329.1 | - | 7 - 8 | 991 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
