Assay Details
Gene Symbol: MX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173940.2 | NP_776365.1 | 9 - 10 | 1191 | 100 | |
GenBank mRNA | AB060169.1 | - | 8 - 9 | 1567 | 100 | |
BC118109.1 | - | 9 - 10 | 1180 | 100 | ||
JQ766265.1 | - | 8 - 9 | 1093 | 100 | ||
U88329.1 | - | 9 - 10 | 1191 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
