Assay Details
Gene Symbol: CAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174004.3 | NP_776429.1 | 1 - 2 | 97 | 72 | |
GenBank mRNA | AY823915.1 | - | 1 - 2 | 97 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_174004.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
