Assay Details
Gene Symbol: IFNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174086.1 | NP_776511.1 | 1 - 2 | 208 | 117 | |
GenBank mRNA | EF675629.1 | - | 1 - 2 | 115 | 117 | |
EU276066.1 | - | 1 - 2 | 138 | 117 | ||
EU277737.1 | - | 1 - 2 | 115 | 117 | ||
FJ263670.1 | - | 1 - 2 | 115 | 117 | ||
M29867.1 | - | 1 - 2 | 208 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
