Assay Details
Gene Symbol: NDUFA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174306.3 | NP_776731.1 | 1 - 2 | 100 | 128 | |
GenBank mRNA | AJ011400.1 | - | 1 - 2 | 102 | 128 | |
BC102502.1 | - | 1 | 104 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
