Assay Details
Gene Symbol: NDUFA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174306.3 | NP_776731.1 | 2 - 3 | 184 | 153 | |
GenBank mRNA | AJ011400.1 | - | 2 - 3 | 186 | 153 | |
BC102502.1 | - | 1 | 188 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
