Assay Details
Gene Symbol: PRDX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174433.2 | NP_776858.1 | 3 - 4 | 572 | 113 | |
GenBank mRNA | AF305563.1 | - | 3 - 4 | 572 | 113 | |
BC109824.1 | - | 3 - 4 | 539 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
