Assay Details
Gene Symbol: SOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174615.2 | NP_777040.1 | 4 - 5 | 430 | 107 | |
GenBank mRNA | AB099022.1 | - | 3 - 4 | 270 | 107 | |
AB099025.1 | - | 3 - 4 | 270 | 107 | ||
BC102432.1 | - | 4 - 5 | 444 | 107 | ||
M81129.1 | - | 4 - 5 | 430 | 107 | ||
MK424005.1 | - | 1 - 2 | 113 | 107 | ||
X54799.1 | - | 348 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
