Assay Details
Gene Symbol: GRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174710.2 | NP_777135.1 | 16 - 17 | 1485 | 80 | |
GenBank mRNA | M34019.1 | - | 16 - 17 | 1485 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
