Assay Details
Gene Symbol: CABP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174728.2 | NP_777153.1 | 2 - 3 | 198 | 70 | |
GenBank mRNA | AF169160.1 | - | 2 - 3 | 198 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
