Assay Details
Gene Symbol: PRDX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174763.2 | NP_777188.1 | 2 - 3 | 431 | 124 | |
GenBank mRNA | AF305562.1 | - | 2 - 3 | 431 | 124 | |
BC102351.1 | - | 3 - 4 | 332 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
