Assay Details
Gene Symbol: MYL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175780.1 | NP_786974.1 | 3 - 4 | 205 | 76 | |
GenBank mRNA | BC103428.1 | - | 3 - 4 | 219 | 76 | |
BC142278.1 | - | 3 - 4 | 192 | 76 | ||
DQ347572.1 | - | 1 - 2 | 87 | 76 | ||
X54977.1 | - | 191 | 76 | |||
X54978.1 | - | 205 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
