Assay Details
Gene Symbol: NDUFS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175810.2 | NP_787004.1 | 2 - 3 | 200 | 78 | |
GenBank mRNA | BC103241.1 | - | 2 - 3 | 197 | 78 | |
X63221.1 | - | 200 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
