Assay Details
Gene Symbol: SDHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175814.3 | NP_787008.1 | 1 | 51 | 79 | |
GenBank mRNA | BC102062.1 | - | 1 | 52 | 79 | |
S74803.1 | - | 1 | 36 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102062.1, NM_175814.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
