Assay Details
Gene Symbol: SDHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175814.3 | NP_787008.1 | 1 | 434 | 73 | |
GenBank mRNA | BC102062.1 | - | 1 | 435 | 73 | |
BT025477.1 | - | 5 - 6 | 404 | 73 | ||
M76747.1 | - | 3 - 4 | 335 | 73 | ||
S74803.1 | - | 1 | 419 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
