Assay Details
Gene Symbol: NDUFA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176658.2 | NP_788831.1 | 2 - 3 | 112 | 98 | |
GenBank mRNA | BC146193.1 | - | 2 - 3 | 110 | 98 | |
X68585.1 | - | 112 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
