Assay Details
Gene Symbol: GSTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177516.1 | NP_803482.1 | 2 - 3 | 112 | 69 | |
GenBank mRNA | BC102704.1 | - | 2 - 3 | 86 | 69 | |
X61233.1 | - | 112 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
