Assay Details
Gene Symbol: EDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181010.2 | NP_851353.1 | 3 - 4 | 683 | 85 | |
GenBank mRNA | BC102459.1 | - | 3 - 4 | 703 | 85 | |
S37093.1 | - | 3 - 4 | 683 | 85 | ||
X52740.1 | - | 433 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
