Assay Details
Gene Symbol: RPL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205797.1 | NP_991366.1 | 1 | 94 | 71 | |
GenBank mRNA | AY528249.1 | - | 1 | 94 | 71 | |
BC102693.1 | - | 1 | 127 | 71 | ||
BC142032.1 | - | 1 | 127 | 71 |
Gene Symbol: LOC784762 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB098966.1 | - | 1 | 94 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
