Assay Details
Gene Symbol: RPL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205797.1 | NP_991366.1 | 1 | 350 | 78 | |
GenBank mRNA | AB098784.1 | - | 3 - 4 | 246 | 78 | |
AY528249.1 | - | 1 | 350 | 78 | ||
BC102693.1 | - | 1 | 383 | 78 | ||
BC142032.1 | - | 1 | 383 | 78 |
Gene Symbol: LOC784762 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB098966.1 | - | 1 | 350 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
