Assay Details
Gene Symbol: HSD11B1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206969.1 | NP_996852.1 | 2 - 3 | 196 | 96 | |
GenBank mRNA | AY554049.1 | - | 2 - 3 | 196 | 96 | |
BC149481.1 | - | 1 - 2 | 480 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC149481.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
