Assay Details
Gene Symbol: HSD11B1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206969.1 | NP_996852.1 | 7 - 8 | 788 | 109 | |
GenBank mRNA | AY554049.1 | - | 7 - 8 | 788 | 109 | |
BC149481.1 | - | 5 - 6 | 960 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
