Assay Details
Gene Symbol: AKR1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012519.1 | NP_001012537.1 | 3 - 4 | 382 | 82 | |
GenBank mRNA | AB098903.1 | - | 3 - 4 | 437 | 82 | |
BC110178.1 | - | 3 - 4 | 439 | 82 | ||
BT021058.1 | - | 3 - 4 | 382 | 82 | ||
M31463.1 | - | 2 - 3 | 298 | 82 | ||
S54973.1 | - | 3 - 4 | 329 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
